Assay Details
| Gene Symbol: IL22RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131220.2 | XP_014986706.2 | 479 | 69 | ||
| XM_015131229.2 | XP_014986715.2 | 478 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL22RA1| Gene Symbol: IL22RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131220.2 | XP_014986706.2 | 479 | 69 | ||
| XM_015131229.2 | XP_014986715.2 | 478 | 69 | |||
Set Membership: |
|