Assay Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257451.1 | NP_001244380.1 | 448 | 118 | ||
| GenBank mRNA | JU336399.1 | - | 448 | 118 | ||
| JU473472.1 | - | 484 | 118 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPAP3
Genomic Map