Assay Details
| Gene Symbol: MAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126679.2 | XP_014982165.2 | 2013 | 60 | ||
| XM_028841114.1 | XP_028696947.1 | 1836 | 60 | |||
| GenBank mRNA | JU486705.1 | - | 411 | 60 | ||
| JV684147.1 | - | 422 | 60 | |||
More Information
Set Membership: |
|

MAF
Genomic Map