Assay Details
| Gene Symbol: DYRK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258143.2 | NP_001245072.1 | 1748 | 58 | ||
| XM_015132883.2 | XP_014988369.1 | 1950 | 58 | |||
| XM_028844778.1 | XP_028700611.1 | 1857 | 58 | |||
| XM_028844779.1 | XP_028700612.1 | 1764 | 58 | |||
| XM_028844780.1 | XP_028700613.1 | 1810 | 58 | |||
| XM_028844781.1 | XP_028700614.1 | 1984 | 58 | |||
| XM_028844782.1 | XP_028700615.1 | 2024 | 58 | |||
| XM_028844783.1 | XP_028700616.1 | 2329 | 58 | |||
| GenBank mRNA | JU333266.1 | - | 1723 | 58 | ||
| JU474571.1 | - | 1723 | 58 | |||
| JU474572.1 | - | 1750 | 58 | |||
| JV043722.1 | - | 1756 | 58 | |||
| JV043723.1 | - | 1729 | 58 | |||
More Information
Set Membership: |
|

DYRK1A
Genomic Map