Assay Details
| Gene Symbol: PDE6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194606.1 | NP_001181535.1 | 331 | 91 | ||
| XM_015111249.2 | XP_014966735.1 | 706 | 91 | |||
| GenBank mRNA | JU334901.1 | - | 220 | 91 | ||
| JU472104.1 | - | 262 | 91 | |||
| JV045081.1 | - | 232 | 91 | |||
| JV547387.1 | - | 258 | 91 | |||
| JV584037.1 | - | 225 | 91 | |||
More Information
Set Membership: |
|

PDE6D
Genomic Map