Assay Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266411.1 | NP_001253340.1 | 310 | 95 | ||
| XM_015140644.2 | XP_014996130.1 | 472 | 95 | |||
| XM_015140645.2 | XP_014996131.1 | 334 | 95 | |||
| GenBank mRNA | JU334139.1 | - | 310 | 95 | ||
More Information
Set Membership: |
|

SNX24
Genomic Map