Assay Details
| Gene Symbol: TK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836864.1 | XP_028692697.1 | 880 | 97 | ||
| GenBank mRNA | JU477000.1 | - | 583 | 97 | ||
| JV660085.1 | - | 66 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TK1
Genomic Map