Assay Details
| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266700.1 | NP_001253629.1 | 687 | 65 | ||
| XM_015139560.2 | XP_014995046.1 | 909 | 65 | |||
| XM_015139561.2 | XP_014995047.1 | 645 | 65 | |||
| GenBank mRNA | JV046777.1 | - | 687 | 65 | ||
More Information
Set Membership: |
|

PDLIM3
Genomic Map