Assay Details
Gene Symbol: CERT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140286.2 | XP_014995772.1 | 1687 | 84 | ||
XM_015140287.1 | XP_014995773.1 | 1533 | 84 | |||
XM_015140288.2 | XP_014995774.1 | 1533 | 84 | |||
XM_028849517.1 | XP_028705350.1 | 1533 | 84 | |||
XM_028849519.1 | XP_028705352.1 | 1533 | 84 | |||
XM_028849520.1 | XP_028705353.1 | 1533 | 84 | |||
GenBank mRNA | JU478502.1 | - | 1369 | 84 | ||
JV704855.1 | - | 1441 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
