Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140286.2 | XP_014995772.1 | 2558 | 116 | ||
| XM_015140288.2 | XP_014995774.1 | 2326 | 116 | |||
| GenBank mRNA | JU478502.1 | - | 2162 | 116 | ||
| JV704855.1 | - | 2234 | 116 | |||
More Information
Set Membership: |
|

CERT1
Genomic Map