Assay Details
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260550.1 | NP_001247479.1 | 486 | 60 | ||
GenBank mRNA | JV043180.1 | - | 486 | 60 | ||
JV656237.1 | - | 303 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
