Assay Details
| Gene Symbol: RNFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109845.4 | XP_001109845.1 | 832 | 122 | ||
| XM_015119990.2 | XP_014975476.1 | 832 | 122 | |||
| XM_015119992.2 | XP_014975478.1 | 832 | 122 | |||
| XM_015119993.2 | XP_014975479.1 | 832 | 122 | |||
| XM_028836254.1 | XP_028692087.1 | 727 | 122 | |||
| GenBank mRNA | JU334804.1 | - | 768 | 122 | ||
| JU475830.1 | - | 731 | 122 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNFT1
Genomic Map