Assay Details
| Gene Symbol: ANKRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266381.1 | NP_001253310.1 | 1037 | 73 | ||
| XM_015140199.2 | XP_014995685.1 | 1409 | 73 | |||
| GenBank mRNA | JU320956.1 | - | 1348 | 73 | ||
| JU476582.1 | - | 1374 | 73 | |||
| JV047780.1 | - | 1037 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANKRA2
Genomic Map