Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109877.3 | XP_001109877.1 | 877 | 85 | ||
| XM_015135351.2 | XP_014990837.1 | 1178 | 85 | |||
| XM_015135353.2 | XP_014990839.1 | 928 | 85 | |||
| GenBank mRNA | JU470856.1 | - | 831 | 85 | ||
More Information
Set Membership: |
|

TEKT2
Genomic Map