Assay Details
| Gene Symbol: CHIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257545.1 | NP_001244474.1 | 517 | 145 | ||
| GenBank mRNA | JU320892.1 | - | 517 | 145 | ||
| JU472879.1 | - | 711 | 145 | |||
| JV045063.1 | - | 767 | 145 | |||
More Information
Set Membership: |
|

CHIC2
Genomic Map