Assay Details
| Gene Symbol: AGPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257833.1 | NP_001244762.1 | 990 | 80 | ||
| XM_015144693.2 | XP_015000179.1 | 1299 | 80 | |||
| XM_028852279.1 | XP_028708112.1 | 1371 | 80 | |||
| XM_028852280.1 | XP_028708113.1 | 3401 | 80 | |||
| XM_028852281.1 | XP_028708114.1 | 865 | 80 | |||
| GenBank mRNA | JU321832.1 | - | 990 | 80 | ||
| JU476275.1 | - | 984 | 80 | |||
| JV669140.1 | - | 1158 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AGPAT5
Genomic Map