Assay Details
| Gene Symbol: NUDT21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126197.2 | XP_014981683.1 | 721 | 143 | ||
| GenBank mRNA | JU320090.1 | - | 690 | 143 | ||
| JU472063.1 | - | 696 | 143 | |||
| JV045091.1 | - | 684 | 143 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NUDT21
Genomic Map