Assay Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138866.2 | XP_014994352.1 | 1075 | 70 | ||
| XM_015138867.2 | XP_014994353.1 | 884 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADH6| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138866.2 | XP_014994352.1 | 1075 | 70 | ||
| XM_015138867.2 | XP_014994353.1 | 884 | 70 | |||
Set Membership: |
|