Assay Details
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130085.2 | XP_014985571.2 | 1309 | 119 | ||
| XM_028844536.1 | XP_028700369.1 | 1309 | 119 | |||
| GenBank mRNA | JU325719.1 | - | 725 | 119 | ||
| JV062333.1 | - | 1155 | 119 | |||
| JV508396.1 | - | 1595 | 119 | |||
More Information
Set Membership: |
|

PODXL2
Genomic Map