Assay Details
| Gene Symbol: ABCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105162.1 | NP_001098632.1 | 71 | 109 | ||
| XM_015135843.2 | XP_014991329.1 | 218 | 109 | |||
| XM_015135844.2 | XP_014991330.1 | 218 | 109 | |||
| XM_015135845.2 | XP_014991331.1 | 218 | 109 | |||
| GenBank mRNA | JU333507.1 | - | 118 | 109 | ||
| JU474776.1 | - | 118 | 109 | |||
| JV046496.1 | - | 117 | 109 | |||
| JV461009.1 | - | 98 | 109 | |||
| JV645802.1 | - | 172 | 109 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ABCF1
Genomic Map