Assay Details
Gene Symbol: LOC114676074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843022.1 | XP_028698855.1 | 1231 | 79 |
Gene Symbol: LAMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001118617.4 | XP_001118617.4 | 3615 | 79 | ||
GenBank mRNA | JV648837.1 | - | 3564 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
