Assay Details
| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090715.4 | XP_001090715.2 | 429 | 108 | ||
| GenBank mRNA | JV713395.1 | - | 142 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HHEX| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090715.4 | XP_001090715.2 | 429 | 108 | ||
| GenBank mRNA | JV713395.1 | - | 142 | 108 | ||
Set Membership: |
|