Assay Details
| Gene Symbol: UCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195393.1 | NP_001182322.1 | 120 | 66 | ||
| XM_015115112.2 | XP_014970598.1 | 951 | 66 | |||
| XM_015115113.2 | XP_014970599.1 | 960 | 66 | |||
| GenBank mRNA | AF202130.1 | - | 112 | 66 | ||
| JU336221.1 | - | 503 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UCP2
Genomic Map