Assay Details
| Gene Symbol: CHST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131917.2 | XP_014987403.1 | 1475 | 101 | ||
| GenBank mRNA | JU477902.1 | - | 526 | 101 | ||
| JV601266.1 | - | 693 | 101 | |||
| JV727282.1 | - | 523 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015131917.2 transcripts.
Set Membership: |
|

CHST2
Genomic Map