Assay Details
| Gene Symbol: LOC717142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116665.2 | XP_014972151.2 | 1103 | 76 | ||
| XM_015116666.2 | XP_014972152.1 | 959 | 76 | |||
| XM_015116667.2 | XP_014972153.1 | 1149 | 76 | |||
| XM_015116668.2 | XP_014972154.1 | 1442 | 76 | |||
| XM_028842759.1 | XP_028698592.1 | 1407 | 76 | |||
| GenBank mRNA | JU473411.1 | - | 816 | 76 | ||
| JV453234.1 | - | 794 | 76 | |||
| JV527769.1 | - | 836 | 76 | |||
| JV568008.1 | - | 872 | 76 | |||
| Gene Symbol: KYAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834738.1 | XP_028690571.1 | 1415 | 76 | ||
| XM_028834739.1 | XP_028690572.1 | 1118 | 76 | |||
| XM_028834740.1 | XP_028690573.1 | 978 | 76 | |||
| XM_028834741.1 | XP_028690574.1 | 1214 | 76 | |||
| XM_028834742.1 | XP_028690575.1 | 863 | 76 | |||
| XM_028834743.1 | XP_028690576.1 | 874 | 76 | |||
| GenBank mRNA | JV601807.1 | - | 868 | 76 | ||
More Information
Set Membership: |
|

LOC717142
Genomic Map