Assay Details
| Gene Symbol: SLC50A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115244.4 | XP_001115244.1 | 404 | 85 | ||
| XM_015111147.2 | XP_014966633.1 | 538 | 85 | |||
| XM_015111155.2 | XP_014966641.1 | 576 | 85 | |||
| XM_015111163.2 | XP_014966649.1 | 261 | 85 | |||
| XM_028828400.1 | XP_028684233.1 | 376 | 85 | |||
| GenBank mRNA | JU321242.1 | - | 367 | 85 | ||
| JU473587.1 | - | 387 | 85 | |||
| JV047654.1 | - | 385 | 85 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC50A1
Genomic Map