Assay Details
| Gene Symbol: PRDM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114407.2 | XP_014969893.2 | 564 | 77 | ||
| XM_015114408.2 | XP_014969894.2 | 766 | 77 | |||
| XM_028833491.1 | XP_028689324.1 | 1586 | 77 | |||
| XM_028833492.1 | XP_028689325.1 | 2036 | 77 | |||
| XM_028833493.1 | XP_028689326.1 | 762 | 77 | |||
| XM_028833494.1 | XP_028689327.1 | 488 | 77 | |||
| XM_028833495.1 | XP_028689328.1 | 261 | 77 | |||
| GenBank mRNA | JV656454.1 | - | 501 | 77 | ||
More Information
Set Membership: |
|

PRDM11
Genomic Map