Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128288.2 | XP_014983774.2 | 2577 | 66 | ||
| XM_015128289.2 | XP_014983775.2 | 2576 | 66 | |||
| XM_015128290.2 | XP_014983776.2 | 2480 | 66 | |||
| XM_028842120.1 | XP_028697953.1 | 2577 | 66 | |||
| GenBank mRNA | JU477214.1 | - | 2386 | 66 | ||
| JV661453.1 | - | 1365 | 66 | |||
More Information
Set Membership: |
|

MAP7D3
Genomic Map