Assay Details
| Gene Symbol: CCBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122113.2 | XP_014977599.2 | 879 | 54 | ||
| XM_015122114.2 | XP_014977600.2 | 584 | 54 | |||
| XM_015122115.2 | XP_014977601.1 | 835 | 54 | |||
More Information
Set Membership: |
|

CCBE1
Genomic Map