Assay Details
| Gene Symbol: LOC106996371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826320.1 | XP_028682153.1 | 1473 | 82 | ||
| XM_028826321.1 | XP_028682154.1 | 803 | 82 | |||
| XM_028826322.1 | XP_028682155.1 | 551 | 82 | |||
| Gene Symbol: LOC709248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826363.1 | XP_028682196.1 | 1511 | 82 | ||
More Information
Set Membership: |
|

LOC106996371
Genomic Map