Assay Details
| Gene Symbol: RHOU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265717.1 | NP_001252646.1 | 323 | 61 | ||
| GenBank mRNA | JU328310.1 | - | 219 | 61 | ||
| JV635798.1 | - | 323 | 61 | |||
More Information
Set Membership: |
|

RHOU
Genomic Map