Assay Details
| Gene Symbol: SOCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028831620.1 | XP_028687453.1 | 215 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028831620.1 transcripts.
Set Membership: |
|

SOCS5
Genomic Map