Assay Details
| Gene Symbol: YES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257512.1 | NP_001244441.1 | 500 | 153 | ||
| XM_015121594.2 | XP_014977080.1 | 385 | 153 | |||
| XM_015121596.2 | XP_014977082.1 | 310 | 153 | |||
| XM_028837690.1 | XP_028693523.1 | 371 | 153 | |||
| XM_028837691.1 | XP_028693524.1 | 462 | 153 | |||
| XM_028837692.1 | XP_028693525.1 | 289 | 153 | |||
| GenBank mRNA | JU319697.1 | - | 500 | 153 | ||
More Information
Set Membership: |
|

YES1
Genomic Map