Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114659.4 | XP_001114659.2 | 1150 | 79 | ||
| XM_028844034.1 | XP_028699867.1 | 1330 | 79 | |||
| GenBank mRNA | JV659078.1 | - | 1033 | 79 | ||
More Information
Set Membership: |
|

CDCP1
Genomic Map