Assay Details
| Gene Symbol: WDR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260815.1 | NP_001247744.1 | 541 | 130 | ||
| XM_015142820.2 | XP_014998306.1 | 616 | 130 | |||
| GenBank mRNA | JU320810.1 | - | 529 | 130 | ||
| JU472217.1 | - | 550 | 130 | |||
| JV045073.1 | - | 541 | 130 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WDR61
Genomic Map