Assay Details
| Gene Symbol: PPIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266543.1 | NP_001253472.1 | 599 | 144 | ||
| XM_015110253.2 | XP_014965739.1 | 864 | 144 | |||
| XM_015110254.2 | XP_014965740.1 | 663 | 144 | |||
| XM_028830538.1 | XP_028686371.1 | 699 | 144 | |||
| XM_028830539.1 | XP_028686372.1 | 939 | 144 | |||
| GenBank mRNA | JU471436.1 | - | 571 | 144 | ||
| JV048725.1 | - | 583 | 144 | |||
| JV633729.1 | - | 626 | 144 | |||
| JV636461.1 | - | 599 | 144 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPIG
Genomic Map