Assay Details
| Gene Symbol: EMSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115228.2 | XP_014970714.1 | 2901 | 62 | ||
| XM_015115229.2 | XP_014970715.1 | 2898 | 62 | |||
| XM_015115230.2 | XP_014970716.1 | 2874 | 62 | |||
| XM_015115231.2 | XP_014970717.1 | 2859 | 62 | |||
| XM_015115232.2 | XP_014970718.1 | 2871 | 62 | |||
| XM_015115233.2 | XP_014970719.1 | 2748 | 62 | |||
| XM_015115234.2 | XP_014970720.1 | 2736 | 62 | |||
| XM_015115236.2 | XP_014970722.1 | 2721 | 62 | |||
| XM_015115237.2 | XP_014970723.1 | 2706 | 62 | |||
| XM_015115239.2 | XP_014970725.1 | 2832 | 62 | |||
| XM_028833834.1 | XP_028689667.1 | 2901 | 62 | |||
| GenBank mRNA | JU335540.1 | - | 2852 | 62 | ||
| JU476467.1 | - | 2852 | 62 | |||
| JV719156.1 | - | 2857 | 62 | |||
More Information
Set Membership: |
|

EMSY
Genomic Map