Assay Details
| Gene Symbol: SRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261334.1 | NP_001248263.1 | 519 | 97 | ||
| GenBank mRNA | JU336064.1 | - | 494 | 97 | ||
| JV044361.1 | - | 519 | 97 | |||
| JV626796.1 | - | 608 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRC
Genomic Map