Assay Details
| Gene Symbol: RFC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266801.1 | NP_001253730.1 | 795 | 114 | ||
| XM_015120869.2 | XP_014976355.1 | 837 | 114 | |||
| XM_028837161.1 | XP_028692994.1 | 824 | 114 | |||
| XM_028837162.1 | XP_028692995.1 | 837 | 114 | |||
| GenBank mRNA | JU335782.1 | - | 750 | 114 | ||
| JU473418.1 | - | 795 | 114 | |||
| JV067800.1 | - | 807 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RFC3
Genomic Map