Assay Details
| Gene Symbol: DSTYK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039949.1 | NP_001035038.1 | 2565 | 66 | ||
| XM_015118269.2 | XP_014973755.1 | 2527 | 66 | |||
| XM_015118274.2 | XP_014973760.1 | 2554 | 66 | |||
| GenBank mRNA | DQ341264.1 | - | 2565 | 66 | ||
| JV636082.1 | - | 2368 | 66 | |||
| JV711422.1 | - | 2419 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DSTYK
Genomic Map