Assay Details
| Gene Symbol: SRFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266668.1 | NP_001253597.1 | 298 | 121 | ||
| XM_015140623.2 | XP_014996109.2 | 494 | 121 | |||
| XM_028849136.1 | XP_028704969.1 | 378 | 121 | |||
| GenBank mRNA | JU333131.1 | - | 303 | 121 | ||
| JU473749.1 | - | 298 | 121 | |||
| JV707939.1 | - | 316 | 121 | |||
More Information
Set Membership: |
|

SRFBP1
Genomic Map