Assay Details
| Gene Symbol: CHORDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257756.1 | NP_001244685.1 | 391 | 92 | ||
| XM_028832509.1 | XP_028688342.1 | 251 | 92 | |||
| GenBank mRNA | JU322407.1 | - | 391 | 92 | ||
| JU322408.1 | - | 391 | 92 | |||
| JU471448.1 | - | 363 | 92 | |||
| JU471449.1 | - | 363 | 92 | |||
| JV043787.1 | - | 514 | 92 | |||
| JV703861.1 | - | 445 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028832509.1 transcripts.
Set Membership: |
|

CHORDC1
Genomic Map