Assay Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194530.1 | NP_001181459.1 | 352 | 62 | ||
| XM_002798178.3 | XP_002798224.1 | 392 | 62 | |||
| XM_015149290.2 | XP_015004776.1 | 262 | 62 | |||
| XM_028827372.1 | XP_028683205.1 | 618 | 62 | |||
| GenBank mRNA | JU336472.1 | - | 200 | 62 | ||
| JV629306.1 | - | 200 | 62 | |||
More Information
Set Membership: |
|

NRSN2
Genomic Map