Assay Details
| Gene Symbol: ABCB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082734.4 | XP_001082734.2 | 1574 | 73 | ||
| GenBank mRNA | JU483501.1 | - | 1123 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCB10| Gene Symbol: ABCB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082734.4 | XP_001082734.2 | 1574 | 73 | ||
| GenBank mRNA | JU483501.1 | - | 1123 | 73 | ||
Set Membership: |
|