Assay Details
| Gene Symbol: HCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140004.2 | XP_014995490.1 | 1629 | 63 | ||
| GenBank mRNA | JV565111.1 | - | 1639 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCN1| Gene Symbol: HCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140004.2 | XP_014995490.1 | 1629 | 63 | ||
| GenBank mRNA | JV565111.1 | - | 1639 | 63 | ||
Set Membership: |
|