Assay Details
| Gene Symbol: MSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257696.1 | NP_001244625.1 | 1447 | 61 | ||
| XM_015127583.2 | XP_014983069.1 | 1286 | 61 | |||
| XM_028841824.1 | XP_028697657.1 | 1512 | 61 | |||
| GenBank mRNA | JU334596.1 | - | 1447 | 61 | ||
| JU470816.1 | - | 1453 | 61 | |||
| JV043599.1 | - | 1455 | 61 | |||
| JV637964.1 | - | 1456 | 61 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MSN
Genomic Map