Assay Details
| Gene Symbol: PRKACB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260959.2 | NP_001247888.1 | 865 | 66 | ||
| XM_015143694.2 | XP_014999180.1 | 883 | 66 | |||
| XM_015143700.2 | XP_014999186.1 | 884 | 66 | |||
| XM_015143702.2 | XP_014999188.1 | 1100 | 66 | |||
| XM_015143707.2 | XP_014999193.1 | 1023 | 66 | |||
| XM_015143711.2 | XP_014999197.1 | 1089 | 66 | |||
| XM_015143717.2 | XP_014999203.1 | 1075 | 66 | |||
| XM_015143732.2 | XP_014999218.1 | 1064 | 66 | |||
| XM_015143740.2 | XP_014999226.1 | 827 | 66 | |||
| XM_015143747.2 | XP_014999233.1 | 1040 | 66 | |||
| XM_015143760.2 | XP_014999246.1 | 883 | 66 | |||
| XM_015143766.2 | XP_014999252.1 | 989 | 66 | |||
| XM_015143768.2 | XP_014999254.1 | 827 | 66 | |||
| XM_028843715.1 | XP_028699548.1 | 1023 | 66 | |||
| GenBank mRNA | JU328231.1 | - | 767 | 66 | ||
| JU335387.1 | - | 868 | 66 | |||
| JU335388.1 | - | 868 | 66 | |||
| JU472629.1 | - | 853 | 66 | |||
| JV043888.1 | - | 867 | 66 | |||
| JV621710.1 | - | 870 | 66 | |||
More Information
Set Membership: |
|

PRKACB
Genomic Map