Assay Details
| Gene Symbol: RNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194648.2 | NP_001181577.1 | 739 | 67 | ||
| XM_015151956.2 | XP_015007442.1 | 896 | 67 | |||
| XM_015151957.2 | XP_015007443.1 | 996 | 67 | |||
| XM_015151960.2 | XP_015007446.1 | 945 | 67 | |||
| XM_015151961.2 | XP_015007447.1 | 764 | 67 | |||
| XM_028828955.1 | XP_028684788.1 | 1012 | 67 | |||
| XM_028828956.1 | XP_028684789.1 | 682 | 67 | |||
| GenBank mRNA | JU332442.1 | - | 866 | 67 | ||
| JU473908.1 | - | 787 | 67 | |||
| JV045863.1 | - | 854 | 67 | |||
| JV601081.1 | - | 861 | 67 | |||
More Information
Set Membership: |
|

RNF41
Genomic Map