Assay Details
| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127137.2 | XP_014982623.1 | 2415 | 87 | ||
| XM_015127139.2 | XP_014982625.1 | 2159 | 87 | |||
| XM_015127140.2 | XP_014982626.1 | 1821 | 87 | |||
| XM_015127141.2 | XP_014982627.1 | 2417 | 87 | |||
| XM_028842401.1 | XP_028698234.1 | 2115 | 87 | |||
| XM_028842402.1 | XP_028698235.1 | 2534 | 87 | |||
| XM_028842403.1 | XP_028698236.1 | 2418 | 87 | |||
| GenBank mRNA | JU333833.1 | - | 2123 | 87 | ||
| JU475042.1 | - | 2076 | 87 | |||
| JV044785.1 | - | 2188 | 87 | |||
More Information
Set Membership: |
|

TAB3
Genomic Map