Assay Details
| Gene Symbol: PAWR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085092.4 | XP_001085092.2 | 1021 | 89 | ||
| XM_015152350.2 | XP_015007836.2 | 1021 | 89 | |||
| GenBank mRNA | JV062375.1 | - | 170 | 89 | ||
| JV652793.1 | - | 427 | 89 | |||
More Information
Set Membership: |
|

PAWR
Genomic Map